4-Chlorothioanisole
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 1-Chloro-4-(methylthio)benzene, 4-Chlorophenyl methyl sulphide
- CAS Number:123-09-1
- Formula:C₇H₇ClS
- MDL Number:MFCD00013643
- Molecular Weight:158.65 g/mol
- Boiling Pt:223 °C
- Density:1.22 g/cm³ (25 °C)
- Flash Pt:90 °C
- Storage Temperature:Refrigerator
- EINECS:204-600-5
3 Options Available Below
Product Details & Documents
Synonyms: 1-Chloro-4-(methylthio)benzene, 4-Chlorophenyl methyl sulphide
- CAS Number:123-09-1
- Formula:C₇H₇ClS
- MDL Number:MFCD00013643
- Molecular Weight:158.65 g/mol
- Boiling Pt:223 °C
- Density:1.22 g/cm³ (25 °C)
- Flash Pt:90 °C
- Storage Temperature:Refrigerator
- EINECS:204-600-5



