Order Entry
Export
ContactUsLinkComponent
2,4-Dibromo-3-nitropyridine 97%
2,4-Dibromo-3-nitropyridine 97%
Catalog # APOSOR345229-250MG
Supplier:  Apollo Scientific
CAS Number:  
2,4-Dibromo-3-nitropyridine 97%
Catalog # APOSOR345229-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :281.8900 g/mol
Storage Temperature:Ambient
CAS Number:121263-10-3
Formula:C₅H₂Br₂N₂O₂
MDL Number:MFCD09750299