Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Chloropyridine-3,4-diamine
5-Chloropyridine-3,4-diamine 97%
5-Chloropyridine-3,4-diamine 97%
APOSOR345157-1G
:
Apollo Scientific
:
5-Chloropyridine-3,4-diamine 97%
APOSOR345157-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
143.57 g/mol
:
Ambient
:
929074-42-0
:
C5H6ClN3
:
MFCD09263519