3',4'-Dihydroxyacetophenone 98%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 1-(3,4-Dihydroxyphenyl)ethan-1-one, Acetopyrocatechol
- CAS Number:1197-09-7
- Formula:C₈H₈O₃
- MDL Number:MFCD00016439
- Molecular Weight:152.15 g/mol
- Storage Temperature:Ambient
5 Options Available Below
Product Details & Documents
Synonyms: 1-(3,4-Dihydroxyphenyl)ethan-1-one, Acetopyrocatechol
- CAS Number:1197-09-7
- Formula:C₈H₈O₃
- MDL Number:MFCD00016439
- Molecular Weight:152.15 g/mol
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



