
3,4-Dimethoxybenzoyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3535-37-3
:
Formula:C₉H₉ClO₃
MDL Number:MFCD00000674
:200.62 g/mol
Boiling Pt:95…98 °C/1mmHg
Melting Pt:70…73 °C
Storage Temperature:Ambient
EINECS:222-568-0
UN:3261
ADR:8,II
:GHS05