Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Diethylphenyl isocyanate
2,6-Diethylphenyl isocyanate 94%
2,6-Diethylphenyl isocyanate 94%
:
Apollo Scientific
2,6-Diethylphenyl isocyanate 94%
:
Apollo Scientific
:
20458-99-5
:
:
C₁₁H₁₃NO
:
MFCD00019912
:
175.23 g/mol
:
2206
:
6.1,II
:
GHS06
:
GHS08
:
Refrigerator