Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl imidazo[1,2-a]pyridine-7-carboxylate
Ethyl imidazo[1,2-a]pyridine-7-carboxylate
Ethyl imidazo[1,2-a]pyridine-7-carboxylate
APOSOR3366-1G
:
Apollo Scientific
:
Ethyl imidazo[1,2-a]pyridine-7-carboxylate
APOSOR3366-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
190.1986 g/mol
:
87…88 °C
:
GHS07
:
Ambient
:
372147-49-4
:
MFCD02325394
:
C₁₀H₁₀N₂O₂
:
MFCD08436046