Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(1,3-Benzoxazol-5-yl)benzaldehyde
4-(1,3-Benzoxazol-5-yl)benzaldehyde 95%
4-(1,3-Benzoxazol-5-yl)benzaldehyde 95%
APOSOR33591-1G
:
Apollo Scientific
:
4-(1,3-Benzoxazol-5-yl)benzaldehyde 95%
APOSOR33591-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
223.23 g/mol
:
Ambient
:
1008361-50-9
:
C₁₄H₉NO₂
:
MFCD18642384