Order Entry
Export
ContactUsLinkComponent
8-Phenyl-1-thia-4-azaspiro[4.5]decan-3-one 95%
8-Phenyl-1-thia-4-azaspiro[4.5]decan-3-one 95%
Catalog # APOSOR33551-1G
Supplier:  Apollo Scientific
CAS Number:  
8-Phenyl-1-thia-4-azaspiro[4.5]decan-3-one 95%
Catalog # APOSOR33551-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :247.36 g/mol
Storage Temperature:Ambient
CAS Number:1221791-99-6
Formula:C₁₄H₁₇NOS
MDL Number:MFCD16140371