1-Thia-4-azaspiro[4.5]decan-6-ol hydrochloride 95%
Catalog # APOSOR33546-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:209.74 g/mol
Storage Temperature:Ambient
CAS Number:1221792-86-4
Formula:C₈H₁₆ClNOS
MDL Number:MFCD16140323