Order Entry
Export
ContactUsLinkComponent
8-Oxa-1-thia-4-azaspiro[4.5]decane hydrochloride 95%
8-Oxa-1-thia-4-azaspiro[4.5]decane hydrochloride 95%
Catalog # APOSOR33540-1G
Supplier:  Apollo Scientific
CAS Number:  
8-Oxa-1-thia-4-azaspiro[4.5]decane hydrochloride 95%
Catalog # APOSOR33540-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :195.71 g/mol
Storage Temperature:Ambient
CAS Number:1221791-86-1
Formula:C₇H₁₄ClNOS
MDL Number:MFCD16140314