Order Entry
Export
ContactUsLinkComponent
(1-Chloro-3,4-dihydro-2-naphthalenyl)methanol 95%
(1-Chloro-3,4-dihydro-2-naphthalenyl)methanol 95%
Catalog # APOSOR33533-1G
Supplier:  Apollo Scientific
CAS Number:  
(1-Chloro-3,4-dihydro-2-naphthalenyl)methanol 95%
Catalog # APOSOR33533-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :194.66 g/mol
Storage Temperature:Ambient
CAS Number:128104-82-5
Formula:C₁₁H₁₁ClO
MDL Number:MFCD12025871