Order Entry
Export
ContactUsLinkComponent
 
1,4-Diazaspiro[4.5]decan-2-one 95%
Catalog # APOSOR33504-1G
Supplier:  Apollo Scientific
CAS Number:  
1,4-Diazaspiro[4.5]decan-2-one 95%
Catalog # APOSOR33504-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :154.21 g/mol
Storage Temperature:Ambient
CAS Number:19718-88-8
Formula:C₈H₁₄N₂O
MDL Number:MFCD14584836