Order Entry
Export
ContactUsLinkComponent
4-(4-Methoxyphenyl)-4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridine 95%
4-(4-Methoxyphenyl)-4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridine 95%
Catalog # APOSOR33404-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(4-Methoxyphenyl)-4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridine 95%
Catalog # APOSOR33404-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :229.28 g/mol
Storage Temperature:Ambient
CAS Number:4875-49-4
Formula:C₁₃H₁₅N₃O
MDL Number:MFCD01839591