Order Entry
Export
ContactUsLinkComponent
4-(3,3-Dimethyl-2-oxo-1-azetanyl)benzenecarbonitrile 95%
4-(3,3-Dimethyl-2-oxo-1-azetanyl)benzenecarbonitrile 95%
Catalog # APOSOR33364-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(3,3-Dimethyl-2-oxo-1-azetanyl)benzenecarbonitrile 95%
Catalog # APOSOR33364-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :200.24 g/mol
Storage Temperature:Ambient
CAS Number:454473-69-9
Formula:C₁₂H₁₂N₂O
MDL Number:MFCD00793440