Order Entry
Export
ContactUsLinkComponent
3-(4-Methoxyphenyl)furan-2,5-dione 95%
3-(4-Methoxyphenyl)furan-2,5-dione 95%
Catalog # APOSOR33113-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(4-Methoxyphenyl)furan-2,5-dione 95%
Catalog # APOSOR33113-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :204.18 g/mol
Storage Temperature:Ambient
CAS Number:4665-00-3
Formula:C₁₁H₈O₄
MDL Number:MFCD00974742