Order Entry
Export
ContactUsLinkComponent
Methyl-3,3-dimethyl-2-(1H-pyrrol-1-yl)butanoate 95%
Methyl-3,3-dimethyl-2-(1H-pyrrol-1-yl)butanoate 95%
Catalog # APOSOR32882-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-3,3-dimethyl-2-(1H-pyrrol-1-yl)butanoate 95%
Catalog # APOSOR32882-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :195.26 g/mol
Storage Temperature:Ambient
CAS Number:476335-37-2
Formula:C₁₁H₁₇NO₂
MDL Number:MFCD01316055