Order Entry
Export
ContactUsLinkComponent
6-(1,4-Thiazinan-4-yl)nicotinaldehyde 95%
6-(1,4-Thiazinan-4-yl)nicotinaldehyde 95%
Catalog # APOSOR32794-1G
Supplier:  Apollo Scientific
CAS Number:  
6-(1,4-Thiazinan-4-yl)nicotinaldehyde 95%
Catalog # APOSOR32794-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :208.28 g/mol
Storage Temperature:Ambient
CAS Number:886360-66-3
Formula:C₁₀H₁₂N₂OS
MDL Number:MFCD04973725