Order Entry
Export
Orders LinkContactUsLinkComponent
3-Hydroxy-2-phenyl-4(1H)-quinolinone 95%

3-Hydroxy-2-phenyl-4(1H)-quinolinone 95%

New Product
Supplier:   Apollo Scientific
3-Hydroxy-2-phenyl-4(1H)-quinolinone 95%
Supplier:  Apollo Scientific
New Product
Certificates
3-Hydroxy-2-phenyl-4(1H)-quinolinone 95%

3 Options Available Below

Product Details & Documents
Synonyms:  3-Hydroxy-2-phenylquinolin-4(1H)-one
  • CAS Number:31588-18-8
  • Formula:C₁₅H₁₁NO₂
  • MDL Number:MFCD00451324
  • Molecular Weight:237.26 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 102,00 €
    102,00 €
    1 * 500 mg
       102,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 163,00 €
    163,00 €
    1 * 1 g
       163,00 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 730,00 €
    730,00 €
    1 * 5 g
       730,00 €