Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
[4-(1H-1,3-Benzimidazol-1-yl)phenyl]methanol
[4-(1H-1,3-Benzimidazol-1-yl)phenyl]methanol 95%
[4-(1H-1,3-Benzimidazol-1-yl)phenyl]methanol 95%
[4-(1H-1,3-Benzimidazol-1-yl)phenyl]methanol 95%
Supplier:
Apollo Scientific
Certificates
Expand image
[4-(1H-1,3-Benzimidazol-1-yl)phenyl]methanol 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
451485-67-9
:
Formula:
C₁₄H₁₂N₂O
MDL Number:
MFCD03425763
:
224.26 g/mol
Storage Temperature:
Ambient