Order Entry
Export
ContactUsLinkComponent
4-(1H-1,3-Benzimidazol-1-yl)benzenecarbaldehyde 95%
4-(1H-1,3-Benzimidazol-1-yl)benzenecarbaldehyde 95%
Catalog # APOSOR32676-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(1H-1,3-Benzimidazol-1-yl)benzenecarbaldehyde 95%
Catalog # APOSOR32676-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :222.25 g/mol
Storage Temperature:Ambient
CAS Number:90514-72-0
Formula:C₁₄H₁₀N₂O
MDL Number:MFCD03425762