Order Entry
Export
ContactUsLinkComponent
5-Methyl[1,6]naphthyridin-2(1H)-one 95%
5-Methyl[1,6]naphthyridin-2(1H)-one 95%
Catalog # APOSOR32646-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Methyl[1,6]naphthyridin-2(1H)-one 95%
Catalog # APOSOR32646-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :160.18 g/mol
Storage Temperature:Ambient
CAS Number:88296-61-1
Formula:C₉H₈N₂O
MDL Number:MFCD00867139