Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,6-Dichloro-N,N-dimethyl-2-pyrimidinamine
4,6-Dichloro-N,N-dimethyl-2-pyrimidinamine 95%
4,6-Dichloro-N,N-dimethyl-2-pyrimidinamine 95%
4,6-Dichloro-N,N-dimethyl-2-pyrimidinamine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
4,6-Dichloro-N,N-dimethyl-2-pyrimidinamine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
5734-68-9
:
Formula:
C₆H₇Cl₂N₃
MDL Number:
MFCD01935962
:
192.05 g/mol
Melting Pt:
56…57.5 °C
Storage Temperature:
Refrigerator