Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5,6-Dihydrobenzo[h]quinazolin-2-amine
5,6-Dihydrobenzo[h]quinazolin-2-amine 95%
5,6-Dihydrobenzo[h]quinazolin-2-amine 95%
5,6-Dihydrobenzo[h]quinazolin-2-amine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
5,6-Dihydrobenzo[h]quinazolin-2-amine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
66521-84-4
:
Formula:
C₁₂H₁₁N₃
MDL Number:
MFCD00139519
:
197.24 g/mol
Storage Temperature:
Ambient