Order Entry
Export
ContactUsLinkComponent
(1E,4E)-1-(Dimethylamino)-5-(2-thienyl)penta-1,4-dien-3-one, Technical Grade
(1E,4E)-1-(Dimethylamino)-5-(2-thienyl)penta-1,4-dien-3-one, Technical Grade
Catalog # APOSOR32375-1G
Supplier:  Apollo Scientific
CAS Number:  
(1E,4E)-1-(Dimethylamino)-5-(2-thienyl)penta-1,4-dien-3-one, Technical Grade
Catalog # APOSOR32375-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :207.3 g/mol
Storage Temperature:Ambient
CAS Number:320416-15-7
Formula:C₁₁H₁₃NOS
MDL Number:MFCD00127282