Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(S)-2-Methylpropane-2-sulphinamide
(S)-2-Methylpropane-2-sulphinamide 98%
(S)-2-Methylpropane-2-sulphinamide 98%
Irritant
(S)-2-Methylpropane-2-sulphinamide 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
(S)-2-Methylpropane-2-sulphinamide 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
343338-28-3
:
Formula:
C₄H₁₁NOS
MDL Number:
MFCD05861480
:
121.20 g/mol
Melting Pt:
104…106 °C
:
GHS07
Storage Temperature:
Refrigerator