1,3-Diethylurea 97%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: N,N'-Diethylurea
- CAS Number:623-76-7
- Formula:C₅H₁₂N₂O
- MDL Number:MFCD00009028
- Molecular Weight:116.16 g/mol
- Melting Pt:109…111 °C
- Storage Temperature:Ambient
5 Options Available Below
Product Details & Documents
Synonyms: N,N'-Diethylurea
- CAS Number:623-76-7
- Formula:C₅H₁₂N₂O
- MDL Number:MFCD00009028
- Molecular Weight:116.16 g/mol
- Melting Pt:109…111 °C
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



