1,4-Dibromobutane
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:110-52-1
:
Formula:BrH₂C(CH₂)₂CH₂Br
MDL Number:MFCD00000261
:215.92 g/mol
Boiling Pt:63…65 °C
Melting Pt:–20 °C
Density:1.808 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:2810
ADR:6.1,III