Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fmoc-11-aminoundecanoic acid
Fmoc-11-aminoundecanoic acid
Fmoc-11-aminoundecanoic acid
Fmoc-11-aminoundecanoic acid
APOSOR322759-5G
:
Apollo Scientific
:
Fmoc-11-aminoundecanoic acid
APOSOR322759-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
423.56 g/mol
:
Ambient
:
88574-07-6
:
C₂₆H₃₃NO₄
:
MFCD00235891