Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Methyl-2,8-diazaspiro[5.5]undecane
2-Methyl-2,8-diazaspiro[5.5]undecane
2-Methyl-2,8-diazaspiro[5.5]undecane
2-Methyl-2,8-diazaspiro[5.5]undecane
Supplier:
Apollo Scientific
Certificates
Expand image
2-Methyl-2,8-diazaspiro[5.5]undecane
Supplier:
Apollo Scientific
About this item
CAS Number:
845290-58-6
:
Formula:
C₁₀H₂₀N₂
MDL Number:
MFCD03768342
:
168.28 g/mol
Storage Temperature:
Ambient