4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine dihydrochloride
Catalog # APOSOR322530-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:196.08 g/mol
Storage Temperature:Ambient
CAS Number:62002-31-7
Formula:C₆H₁₁Cl₂N₃
MDL Number:MFCD00182087