2,3-Dihydrothieno[3,4-b]-1,4-dioxin-5-carbaldehyde
Catalog # APOSOR322518-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:170.19 g/mol
Storage Temperature:Ambient
CAS Number:204905-77-1
Formula:C₇H₆O₃S
MDL Number:MFCD01651766