Order Entry
Export
Orders LinkContactUsLinkComponent
1,4-Oxazepane

1,4-Oxazepane

New Product
Supplier:   Apollo Scientific
1,4-Oxazepane
Supplier:  Apollo Scientific
New Product
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  Homomorpholine
  • CAS Number:5638-60-8
  • Formula:C₅H₁₁NO
  • MDL Number:MFCD09971804
  • Molecular Weight:101.1 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 65,20 €
    65,20 €
    1 * 1 g
       65,20 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 316,00 €
    316,00 €
    1 * 5 g
       316,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 530,00 €
    530,00 €
    1 * 25 g
       530,00 €

    Specifications:
    New Product
    New Product