Order Entry
Export
ContactUsLinkComponent
Methyl dimethoxyacetate
Methyl dimethoxyacetate
Catalog # APOSOR322201-5G
Supplier:  Apollo Scientific
CAS Number:  
Methyl dimethoxyacetate
Catalog # APOSOR322201-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :134.13 g/mol
Storage Temperature:Ambient
CAS Number:89-91-8
Formula:C₅H₁₀O₄
MDL Number:MFCD00008484