Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,1-Dibromoformaldoxime
1,1-Dibromoformaldoxime 95%
1,1-Dibromoformaldoxime 95%
Corrosive
Irritant
1,1-Dibromoformaldoxime 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
Irritant
1,1-Dibromoformaldoxime 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
74213-24-4
:
Formula:
CHBr₂NO
Melting Pt:
71…73 °C
:
GHS05
:
GHS07
Storage Temperature:
Ambient