3-(3,4-Methylenedioxyphenyl)propionic acid
Catalog # APOSOR322185-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:194.19 g/mol
Storage Temperature:Ambient
CAS Number:2815-95-4
Formula:C₁₀H₁₀O₄
MDL Number:MFCD00016909