Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Dichloroisoquinoline
1,3-Dichloroisoquinoline
1,3-Dichloroisoquinoline
Irritant
1,3-Dichloroisoquinoline
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1,3-Dichloroisoquinoline
Supplier:
Apollo Scientific
About this item
CAS Number:
7742-73-6
:
Formula:
C₉H₅Cl₂N
MDL Number:
MFCD00034750
:
198.05 g/mol
Melting Pt:
121…122 °C
:
GHS07
Storage Temperature:
Ambient