N-Methyl-1,3-propanediamine 98%
Catalog # APOSOR322100-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:88.15 g/mol
Storage Temperature:Ambient
CAS Number:6291-84-5
Formula:C₄H₁₂N₂
MDL Number:MFCD00008209