Diethyl pyrazole-3,5-dicarboxylate
Catalog # APOSOR322084-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:212.21 g/mol
Storage Temperature:Ambient
CAS Number:37687-24-4
Formula:C₉H₁₂N₂O₄
MDL Number:MFCD00152167