Order Entry
Export
ContactUsLinkComponent
1,8-Dinitro-9,10-anthraquinone 95%
Irritant
1,8-Dinitro-9,10-anthraquinone 95%
Catalog # APOSOR322049-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,8-Dinitro-9,10-anthraquinone 95%
Catalog # APOSOR322049-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :298.21
 :GHS07
Storage Temperature:Ambient
CAS Number:129-39-5
Formula:C₁₄H₆N₂O₆