
1,2-Diamino-9,10-anthraquinone
Catalog # APOSOR322042-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Melting Pt:288 °C
:238.241 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1758-68-5
Formula:C₁₄H₁₀N₂O₂
MDL Number:MFCD00001219