

3-Formyl-2,6-dimethoxypyridine 97%
Catalog # APOSOR3215-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:167.17 g/mol
Melting Pt:68…72 °C
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:58819-72-0
Formula:C₈H₉NO₃
MDL Number:MFCD08064048