0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
1-Benzyl-3-methylpiperazine-1,3-dicarboxylate
1-Benzyl-3-methylpiperazine-1,3-dicarboxylate 97%
1-Benzyl-3-methylpiperazine-1,3-dicarboxylate 97%
APOSOR321353-1G
:
Apollo Scientific
:
1-Benzyl-3-methylpiperazine-1,3-dicarboxylate 97%
APOSOR321353-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
278.3037 g/mol
:
GHS07
:
Ambient
:
129799-11-7
:
C₁₄H₁₈N₂O₄
:
MFCD03001724