Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1H-Pyrrolo[3,2-c]pyridine-3-carboxaldehyde
1H-Pyrrolo[3,2-c]pyridine-3-carboxaldehyde 97%
1H-Pyrrolo[3,2-c]pyridine-3-carboxaldehyde 97%
:
Apollo Scientific
1H-Pyrrolo[3,2-c]pyridine-3-carboxaldehyde 97%
:
Apollo Scientific
:
933717-10-3
:
:
C₈H₆N₂O
:
MFCD08690128
:
146.15 g/mol
:
GHS07
:
Ambient