
8-Bromo-1,6-naphthyridin-5(6H)-one
Catalog # APOSOR318080-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:225.05 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:155057-97-9
Formula:C₈H₅BrN₂O
MDL Number:MFCD00928592