Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(4-Acetyl-1,4-diazepan-1-yl)ethanamine
2-(4-Acetyl-1,4-diazepan-1-yl)ethanamine 95+%
2-(4-Acetyl-1,4-diazepan-1-yl)ethanamine 95+%
2-(4-Acetyl-1,4-diazepan-1-yl)ethanamine 95+%
Supplier:
Apollo Scientific
Certificates
Expand image
2-(4-Acetyl-1,4-diazepan-1-yl)ethanamine 95+%
Supplier:
Apollo Scientific
About this item
CAS Number:
928003-92-3
:
Formula:
C₉H₁₉N₃O
MDL Number:
MFCD09258788
:
185.27 g/mol
Storage Temperature:
Ambient