
5-Iodo-m-xylene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:22445-41-6
:
Formula:C₈H₉I
MDL Number:MFCD00060659
:232.06 g/mol
Boiling Pt:92…94 °C/3mmHg
Density:1.608 g/cm³ (25 °C)
Flash Pt:113 °C
:GHS07
Storage Temperature:Ambient