Diisopropyl-3,3-dimethoxycyclobutane-1,1-dicarboxylate
Catalog # APOSOR317057-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:288.34 g/mol
Storage Temperature:Ambient
CAS Number:115118-68-8
Formula:C₁₄H₂₄O₆
MDL Number:MFCD06742779