Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7H-Pyrrolo[2,3-d]pyrimidine
7H-Pyrrolo[2,3-d]pyrimidine
7H-Pyrrolo[2,3-d]pyrimidine
Irritant
7H-Pyrrolo[2,3-d]pyrimidine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
7H-Pyrrolo[2,3-d]pyrimidine
Supplier:
Apollo Scientific
Certificates
About this item
CAS Number:
271-70-5
:
Formula:
C₆H₅N₃
MDL Number:
MFCD08234403
Melting Pt:
130…133 °C
:
119.1240 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
Documentation