0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
4,4-Dimethyl-1,3-cyclohexanedione
4,4-Dimethyl-1,3-cyclohexanedione
4,4-Dimethyl-1,3-cyclohexanedione
4,4-Dimethyl-1,3-cyclohexanedione
Supplier:
Apollo Scientific
Certificates
Expand image
4,4-Dimethyl-1,3-cyclohexanedione
Supplier:
Apollo Scientific
About this item
CAS Number:
562-46-9
:
Formula:
C₈H₁₂O₂
MDL Number:
MFCD00075224
:
140.18 g/mol
Boiling Pt:
130 °C/7mmHg
Melting Pt:
102…105 °C
Storage Temperature:
Ambient